Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zmynd12
Entrez Gene ID
313552
Gene Name
zinc finger, MYND-type containing 12
Gene Aliases
-
Chromosome Location
Chr.5: 138470173 - 138505928 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107970.1 NP_001101440.1 2 - 3 311 80

View Full Product Details