Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krt79
Entrez Gene ID
683720
Gene Name
keratin 79, type II
Gene Aliases
-
Chromosome Location
Chr.7: 143528204 - 143538777 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750410.3 XP_003750458.1 4 - 5 915 72

View Full Product Details