Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt79
Entrez Gene ID
683720
Gene Name
keratin 79, type II
Gene Aliases
-
Chromosome Location
Chr.7: 143528204 - 143538777 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750410.3 XP_003750458.1 8 - 9 1459 72

View Full Product Details