Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krt82
Entrez Gene ID
366991
Gene Name
keratin 82, type II
Gene Aliases
Kb22
Chromosome Location
Chr.7: 143196282 - 143206138 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008803.1 NP_001008803.1 1 - 2 336 80

View Full Product Details