The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106198.1 | NP_001099668.1 | 6 - 7 | 1330 | 69 |
XM_006255807.2 | XP_006255869.1 | 6 - 7 | 1417 | 69 |