Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcnxl2
Entrez Gene ID
307949
Gene Name
pecanex-like 2 (Drosophila)
Gene Aliases
RGD1305883
Chromosome Location
Chr.19: 58583000 - 58736065 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006255821.2 XP_006255883.1 17 - 18 4119 131

View Full Product Details