Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rbm34
Entrez Gene ID
307956
Gene Name
RNA binding motif protein 34
Gene Aliases
RGD1310161
Chromosome Location
Chr.19: 59874745 - 59894937 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014015.1 NP_001014037.1 1 - 2 64 62
GenBank mRNA BC087155.1 - 1 - 2 64 62

View Full Product Details