Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plcxd3
Entrez Gene ID
310358
Gene Name
phosphatidylinositol-specific phospholipase C, X domain containing 3
Gene Aliases
RGD1308448
Chromosome Location
Chr.2: 54191538 - 54360780 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107671.1 NP_001101141.1 2 - 3 1040 58

View Full Product Details