Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ranbp3l
Entrez Gene ID
294789
Gene Name
RAN binding protein 3-like
Gene Aliases
RGD1307559
Chromosome Location
Chr.2: 58221457 - 58235333 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006232030.2 XP_006232092.1 - 743 81

View Full Product Details