Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Calr
Entrez Gene ID
64202
Gene Name
calreticulin
Gene Aliases
-
Chromosome Location
Chr.19: 25956771 - 25961666 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022399.2 NP_071794.1 4 - 5 550 76
GenBank mRNA BC062395.1 - 4 - 5 527 76
BU671467.1 - 4 - 5 550 76
D78308.1 - 4 - 5 502 76
FQ210587.1 - - 555 76
FQ220096.1 - 4 - 5 556 76
X53363.1 - 4 - 5 522 76
X79327.1 - 4 - 5 500 76

View Full Product Details