The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107163.1 | NP_001100633.1 | 11 - 12 | 2242 | 55 |
XM_006255251.2 | XP_006255313.1 | 11 - 12 | 2281 | 55 |