Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
E2f4
Entrez Gene ID
100360427
Gene Name
E2F transcription factor 4, p107/p130-binding
Gene Aliases
-
Chromosome Location
Chr.19: 37252843 - 37260239 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271345.1 NP_001258274.1 4 - 5 539 66
XM_006255433.1 XP_006255495.1 4 - 5 534 66

View Full Product Details