Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Usp38
Entrez Gene ID
307764
Gene Name
ubiquitin specific peptidase 38
Gene Aliases
-
Chromosome Location
Chr.19: 30668269 - 30699852 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751862.3 XP_003751910.1 9 - 10 3497 64

View Full Product Details