The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042621.1 | NP_001036086.1 | 12 - 13 | 1785 | 65 |
XM_008761315.1 | XP_008759537.1 | 12 - 13 | 1914 | 65 | |
XM_008761316.1 | XP_008759538.1 | 12 - 13 | 1911 | 65 | |
GenBank mRNA | AY724476.1 | - | 12 - 13 | 1785 | 65 |