Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmod4
Entrez Gene ID
295261
Gene Name
tropomodulin 4
Gene Aliases
-
Chromosome Location
Chr.2: 196295069 - 196299896 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106449.2 NP_001099919.1 5 - 6 650 78
NM_001270650.1 NP_001257579.1 6 - 7 724 78
NM_001270651.1 NP_001257580.1 6 - 7 697 78
NM_001270652.1 NP_001257581.1 5 - 6 627 78
NM_001270653.1 NP_001257582.1 5 - 6 654 78
XM_006232855.2 XP_006232917.1 6 - 7 719 78
XM_008761282.1 XP_008759504.1 5 - 6 685 78
GenBank mRNA FM111895.1 - 2 - 3 106 78
FQ214903.1 - 5 - 6 587 78
FQ215988.1 - 6 - 7 653 78
FQ216125.1 - 5 - 6 576 78
FQ216236.1 - 3 - 4 321 78
FQ216249.1 - 6 - 7 647 78
FQ216534.1 - 5 - 6 553 78
FQ217203.1 - 5 - 6 574 78
FQ224853.1 - 5 - 6 575 78

View Full Product Details