Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddx20
Entrez Gene ID
84473
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 20
Gene Aliases
Dp103
Chromosome Location
Chr.2: 208144142 - 208154805 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191711.1 NP_001178640.1 3 - 4 565 120
XM_006233099.2 XP_006233161.1 4 - 5 3071 120

View Full Product Details