Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hist2h4
Entrez Gene ID
295277
Gene Name
histone cluster 2, H4
Gene Aliases
Hist4h4
Chromosome Location
Chr.2: 198411955 - 198412350 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123469.1 NP_001116941.1 1 - 1 335 108

View Full Product Details