The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009635.1 | NP_001009635.1 | 4 - 5 | 582 | 95 |
GenBank mRNA | BC088471.1 | - | 4 - 5 | 582 | 95 |