The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107861.1 | NP_001101331.1 | 3 - 4 | 718 | 81 |
XM_006236547.2 | XP_006236609.1 | 3 - 4 | 1029 | 81 | |
XM_006236548.2 | XP_006236610.1 | 3 - 4 | 873 | 81 | |
XM_006236549.2 | XP_006236611.1 | 3 - 4 | 1290 | 81 | |
XM_008762892.1 | XP_008761114.1 | 3 - 4 | 1395 | 81 | |
XM_008762893.1 | XP_008761115.1 | 4 - 5 | 988 | 81 | |
XM_008762894.1 | XP_008761116.1 | 3 - 4 | 763 | 81 |