Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Znrf2
Entrez Gene ID
362367
Gene Name
zinc and ring finger 2
Gene Aliases
Znrf1
Chromosome Location
Chr.4: 85009350 - 85090914 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108628.1 NP_001102098.1 4 - 5 777 125

View Full Product Details