Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gimap9
Entrez Gene ID
493865
Gene Name
GTPase, IMAP family member 9
Gene Aliases
-
Chromosome Location
Chr.4: 78310379 - 78313212 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008398.2 NP_001008399.1 1 - 2 49 74
GenBank mRNA BC103635.1 - 1 - 2 49 74
DQ125337.1 - 1 - 2 48 74
DQ125338.1 - 1 - 2 48 74

View Full Product Details