Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atp6v1a
Entrez Gene ID
685232
Gene Name
ATPase, H+ transporting, lysosomal V1 subunit A
Gene Aliases
Atp6v1a1
Chromosome Location
Chr.11: 61531391 - 61584634 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108318.2 NP_001101788.1 5 - 6 664 65
XM_006248348.2 XP_006248410.1 5 - 6 865 65
XM_006248349.2 XP_006248411.1 5 - 6 856 65
GenBank mRNA FQ211993.1 - 5 - 6 697 65

View Full Product Details