Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arl6
Entrez Gene ID
363760
Gene Name
ADP-ribosylation factor-like 6
Gene Aliases
-
Chromosome Location
Chr.11: 42858467 - 42884324 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006248193.2 XP_006248255.1 7 - 8 1075 92
GenBank mRNA BC162029.1 - 7 - 8 922 92

View Full Product Details