Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ift57
Entrez Gene ID
303968
Gene Name
intraflagellar transport 57
Gene Aliases
Esrrbl1
Chromosome Location
Chr.11: 53664638 - 53731779 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107093.1 NP_001100563.1 8 - 9 1026 109

View Full Product Details