Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zdhhc19
Entrez Gene ID
288045
Gene Name
zinc finger, DHHC-type containing 19
Gene Aliases
RGD1560310
Chromosome Location
Chr.11: 71490346 - 71522952 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039259.1 NP_001034348.1 3 - 4 612 77
XM_006248447.2 XP_006248509.1 3 - 4 614 77
XM_008768710.1 XP_008766932.1 3 - 4 620 77
GenBank mRNA AY886535.1 - 3 - 4 612 77
BC127441.1 - 3 - 4 614 77

View Full Product Details