The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039259.1 | NP_001034348.1 | 4 - 5 | 745 | 65 |
XM_006248447.2 | XP_006248509.1 | 4 - 5 | 747 | 65 | |
XM_008768710.1 | XP_008766932.1 | 4 - 5 | 753 | 65 | |
GenBank mRNA | AY886535.1 | - | 4 - 5 | 745 | 65 |
BC127441.1 | - | 4 - 5 | 747 | 65 |