The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025015.1 | NP_001020186.1 | 3 - 4 | 582 | 142 |
XM_006248340.2 | XP_006248402.1 | 2 - 3 | 508 | 142 | |
GenBank mRNA | BC091236.1 | - | 3 - 4 | 582 | 142 |
FQ216550.1 | - | 3 - 4 | 142 |