Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gtpbp8
Entrez Gene ID
360714
Gene Name
GTP-binding protein 8 (putative)
Gene Aliases
RGD1310025
Chromosome Location
Chr.11: 60906990 - 60916958 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025015.1 NP_001020186.1 1 - 2 357 93
GenBank mRNA BC091236.1 - 1 - 2 357 93
FQ216550.1 - 1 - 2 356 93

View Full Product Details