Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam3b
Entrez Gene ID
304050
Gene Name
family with sequence similarity 3, member B
Gene Aliases
RGD1311662
Chromosome Location
Chr.11: 37922308 - 37993270 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107102.1 NP_001100572.1 1 - 2 111 90
XM_006248157.2 XP_006248219.1 1 - 2 174 90

View Full Product Details