Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Irgc
Entrez Gene ID
308428
Gene Name
immunity-related GTPase family, cinema
Gene Aliases
Ifgge, Irgc1, RGD1311107
Chromosome Location
Chr.1: 81291743 - 81295517 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014016.1 NP_001014038.1 1 - 2 173 104
GenBank mRNA BC079061.1 - 1 - 2 173 104

View Full Product Details