Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dkkl1
Entrez Gene ID
499150
Gene Name
dickkopf-like 1
Gene Aliases
RGD1563523
Chromosome Location
Chr.1: 101205426 - 101210675 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109145.1 NP_001102615.1 2 - 3 529 107

View Full Product Details