Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rinl
Entrez Gene ID
292760
Gene Name
Ras and Rab interactor-like
Gene Aliases
RGD1310133
Chromosome Location
Chr.1: 86972004 - 86983306 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106241.1 NP_001099711.1 5 - 6 496 60
XM_006228653.2 XP_006228715.1 5 - 6 748 60
XM_006228654.2 XP_006228716.1 5 - 6 736 60
XM_006228655.2 XP_006228717.1 5 - 6 748 60
XM_006228656.2 XP_006228718.1 5 - 6 748 60
XM_006228657.2 XP_006228719.1 4 - 5 471 60
XM_008759120.1 XP_008757342.1 4 - 5 613 60
XM_008759121.1 XP_008757343.1 5 - 6 581 60

View Full Product Details