Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcaf10
Entrez Gene ID
313242
Gene Name
DDB1 and CUL4 associated factor 10
Gene Aliases
RGD1304730, Wdr32
Chromosome Location
Chr.5: 60900414 - 60937547 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107935.1 NP_001101405.1 2 - 3 694 82

View Full Product Details