The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108972.2 | NP_001102442.1 | 3 - 4 | 845 | 64 |
GenBank mRNA | BC158647.1 | - | 3 - 4 | 845 | 64 |