Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zdhhc21
Entrez Gene ID
298184
Gene Name
zinc finger, DHHC-type containing 21
Gene Aliases
-
Chromosome Location
Chr.5: 100924621 - 100983380 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039009.1 NP_001034098.1 5 - 6 697 88
XM_008763765.1 XP_008761987.1 5 - 6 3599 88
XM_008763766.1 XP_008761988.1 6 - 7 895 88
XM_008763767.1 XP_008761989.1 6 - 7 754 88
XM_008763768.1 XP_008761990.1 6 - 7 3308 88
GenBank mRNA AY886536.1 - 5 - 6 697 88

View Full Product Details