Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh3gl2
Entrez Gene ID
116743
Gene Name
SH3-domain GRB2-like 2
Gene Aliases
Sh3d2a, Sh3p4
Chromosome Location
Chr.5: 103479767 - 103654507 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053935.1 NP_446387.1 4 - 5 420 55
XM_006238359.2 XP_006238421.1 4 - 5 1804 55

View Full Product Details