The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053935.1 | NP_446387.1 | 7 - 8 | 817 | 66 |
XM_006238359.2 | XP_006238421.1 | 7 - 8 | 2201 | 66 | |
GenBank mRNA | AF009603.1 | - | 4 - 5 | 419 | 66 |