Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ano7
Entrez Gene ID
367318
Gene Name
anoctamin 7
Gene Aliases
Ngep, Tmem16g
Chromosome Location
Chr.9: 100509660 - 100553261 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004071.1 NP_001004071.1 19 - 20 2109 85
XM_008767382.1 XP_008765604.1 20 - 21 3763 85

View Full Product Details