Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vapa
Entrez Gene ID
58857
Gene Name
VAMP (vesicle-associated membrane protein)-associated protein A
Gene Aliases
-
Chromosome Location
Chr.9: 113300831 - 113331872 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008767384.1 XP_008765606.1 5 - 6 1322 64

View Full Product Details