The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013179.1 | NP_001013197.1 | 1 - 2 | 329 | 57 |
XM_006245407.2 | XP_006245469.1 | 2 - 3 | 282 | 57 | |
GenBank mRNA | BC087597.1 | - | 1 - 2 | 329 | 57 |