Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hes6
Entrez Gene ID
316626
Gene Name
hes family bHLH transcription factor 6
Gene Aliases
-
Chromosome Location
Chr.9: 98549698 - 98551439 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013179.1 NP_001013197.1 2 - 3 412 116
XM_006245407.2 XP_006245469.1 3 - 4 365 116
GenBank mRNA BC087597.1 - 2 - 3 412 116

View Full Product Details