The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175846.2 | NP_787040.2 | 1 - 2 | 856 | 78 |
GenBank mRNA | AF461735.1 | - | 1 - 2 | 859 | 78 |
AY435135.1 | - | 1 - 2 | 856 | 78 |