The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031064.2 | NP_112326.1 | 7 - 8 | 712 | 60 |
GenBank mRNA | BC058495.1 | - | 7 - 8 | 716 | 60 |
FQ215131.1 | - | 4 - 5 | 385 | 60 | |
M22756.1 | - | 7 - 8 | 637 | 60 |