Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stam
Entrez Gene ID
498798
Gene Name
signal transducing adaptor molecule (SH3 domain and ITAM motif) 1
Gene Aliases
RGD1564499
Chromosome Location
Chr.17: 81209867 - 81255808 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109121.2 NP_001102591.1 7 - 8 846 63
GenBank mRNA BC168933.1 - 7 - 8 846 63
FQ221736.1 - 7 - 8 923 63

View Full Product Details