Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akr1cl
Entrez Gene ID
361267
Gene Name
aldo-keto reductase family 1, member C-like
Gene Aliases
Akr1cl1
Chromosome Location
Chr.17: 69835338 - 69862093 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109900.1 NP_001103370.1 1 - 2 132 78

View Full Product Details