Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St8sia6
Entrez Gene ID
291325
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 6
Gene Aliases
Siat8f
Chromosome Location
Chr.17: 80960785 - 81002830 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213624.1 NP_998789.1 - 637 76
GenBank mRNA AJ699423.1 - - 637 76

View Full Product Details