Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Reep1
Entrez Gene ID
362384
Gene Name
receptor accessory protein 1
Gene Aliases
RGD1305230
Chromosome Location
Chr.4: 99618537 - 99735329 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108633.1 NP_001102103.1 3 - 4 323 81
XM_008762991.1 XP_008761213.1 4 - 5 292 81
XM_008762992.1 XP_008761214.1 3 - 4 364 81
XM_008762993.1 XP_008761215.1 4 - 5 292 81
XM_008762994.1 XP_008761216.1 3 - 4 408 81
GenBank mRNA FQ217879.1 - 3 - 4 281 81

View Full Product Details