Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smyd1
Entrez Gene ID
297333
Gene Name
SET and MYND domain containing 1
Gene Aliases
-
Chromosome Location
Chr.4: 99074535 - 99125142 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106595.1 NP_001100065.1 6 - 7 937 111
XM_006236632.2 XP_006236694.1 5 - 6 929 111

View Full Product Details