Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp6v1b1
Entrez Gene ID
312488
Gene Name
ATPase, H transporting, lysosomal V1 subunit B1
Gene Aliases
-
Chromosome Location
Chr.4: 115417100 - 115435754 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107867.2 NP_001101337.2 5 - 6 505 72

View Full Product Details