Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snrpg
Entrez Gene ID
681031
Gene Name
small nuclear ribonucleoprotein polypeptide G
Gene Aliases
-
Chromosome Location
Chr.4: 118167294 - 118174843 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135085.1 NP_001128557.1 2 - 3 131 87
GenBank mRNA BC168750.1 - 2 - 3 97 87
BI285268.1 - 2 - 3 87
FQ217038.1 - 2 - 3 97 87
FQ221105.1 - 2 - 3 129 87
FQ221517.1 - 2 - 3 97 87
FQ221580.1 - 2 - 3 158 87
FQ221827.1 - 2 - 3 99 87
FQ222397.1 - 2 - 3 97 87
FQ222427.1 - 2 - 3 100 87
FQ222653.1 - 2 - 3 146 87
FQ223239.1 - 2 - 3 97 87
FQ223409.1 - 2 - 3 146 87
FQ228483.1 - 2 - 3 100 87
FQ228534.1 - 2 - 3 97 87
FQ228634.1 - 2 - 3 105 87
FQ228635.1 - 2 - 3 142 87

Gene Symbol
LOC687679
Entrez Gene ID
687679
Gene Name
similar to small nuclear ribonucleoprotein polypeptide G
Gene Aliases
-
Chromosome Location
Chr.1: 91588215 - 91588610 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003748839.2 XP_003748887.1 1 - 1 98 87

View Full Product Details